Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,6-diphenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A29211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29211G 1 g
A29215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29211G Supplier TCI America Supplier No. A29211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40230-24-8
Molecular Formula C16H13N3
Molecular Weight (g/mol) 247.301
MDL Number MFCD00234910
InChI Key KZUCBEYDRUCBCS-UHFFFAOYSA-N
PubChem CID 619024
IUPAC Name 4,6-diphenylpyrimidin-2-amine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)N)C3=CC=CC=C3

Specifications

Melting Point 136°C
Quantity 1 g
Formula Weight 247.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.