Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-bromopyridine 98.0+%, TCI America™
SDP

Catalog No. A23391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23391G 1 g
A23395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23391G Supplier TCI America Supplier No. A23391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84249-14-9
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD01646115
InChI Key BAQKUNMKVAPWGU-UHFFFAOYSA-N
Synonym 2-amino-4-bromopyridine, 2-pyridinamine, 4-bromo, 4-bromo-2-pyridinamine, 4-bromopyridin-2-ylamine, 4-bromo-2-pyridylamine, 4-bromo-2-amine-pyridine, 4-bromo-pyridin-2-ylamine, 4-bromo-2-aminopyridine, 4-brompyridin-2-amin, pubchem4039
PubChem CID 693282
IUPAC Name 4-bromopyridin-2-amine
SMILES C1=CN=C(C=C1Br)N

Specifications

Melting Point 147°C
Quantity 1 g
Formula Weight 173.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-bromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.