Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloro-6-methylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A108825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A108825G 25 g
A10885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A108825G Supplier TCI America Supplier No. A108825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5600-21-5
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD00006091
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methylpyrimidine, am inhibitor, am pesticide, prepn. am, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, 2-pyrimidinamine, 4-chloro-6-methyl, am nitrification inhibitor, pyrimidine, 2-amino-4-chloro-6-methyl, 2-amino-6-chloro-4-methylpyrimidine
PubChem CID 21810
ChEBI CHEBI:58960
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)Cl

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 25 g
Formula Weight 143.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine
EINECSNumber (5)-0928
RTECSNumber UV6329600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.