Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-cyanopyridine, 97%

Catalog No. AAL1984106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1984106 5 g
AAL1984103 1 g
AAL1984114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1984106 Supplier Thermo Scientific Chemicals Supplier No. L1984106
Add to Cart

CAS: 42182-27-4 | C6H5N3 | 119.127 g/mol

2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42182-27-4
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD03791310
InChI Key GEEAYLFEIFJFGP-UHFFFAOYSA-N
Synonym 2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine
PubChem CID 7015524
IUPAC Name 2-aminopyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)N

Specifications

Melting Point 148°C to 152°C
Quantity 5 g
UN Number UN3439
Beilstein 386393
Solubility Information Slightly soluble in water.
Formula Weight 119.13
Percent Purity 97%
Chemical Name or Material 2-Amino-4-cyanopyridine
Hazard Category H301+H311-H319
Hazard Statement GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.