Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methoxy-6-methylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A0950500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0950500G 500 g
A095025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0950500G Supplier TCI America Supplier No. A0950500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7749-47-5
Molecular Formula C6H9N3O
Molecular Weight (g/mol) 139.158
MDL Number MFCD00059768
InChI Key SNWZXTZIZWBIDQ-UHFFFAOYSA-N
Synonym 2-amino-4-methoxy-6-methylpyrimidine, 2-amino-4-methyl-6-methoxypyrimidine, 2-pyrimidinamine, 4-methoxy-6-methyl, 4-methoxy-6-methyl-2-pyrimidinamine, 4-methoxy-6-methylpyrimidine-2-ylamine, pubchem21464, acmc-209paa, pyrimidine, 2-amino-4-methoxy-6-methyl, ksc496m2l, 2amino-4-methyl-6-methoxypyrimidine
PubChem CID 587236
IUPAC Name 4-methoxy-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)OC

Specifications

Melting Point 158°C
Color White
Quantity 500 g
Formula Weight 139.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxy-6-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.