Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methyl-5-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAL1972906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972906 5 g
AAL1972903 1 g
AAL1972914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1972906 Supplier Thermo Scientific Chemicals Supplier No. L1972906
Add to Cart

2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.

Solubility
Soluble in DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21901-40-6
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00010692
InChI Key GRBBNZYMXKTQAI-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
PubChem CID 243165
IUPAC Name 4-methyl-5-nitropyridin-2-amine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])N

Specifications

Melting Point 223°C to 225°C
Quantity 5 g
Beilstein 137695
Solubility Information Soluble in DMSO.
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methyl-5-nitropyridine
Hazard Category H315-H319-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.