Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylthiazole Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A033625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033625G 25 g
A03365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A033625G Supplier TCI America Supplier No. A033625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6142-15-0
Molecular Formula C4H7ClN2S
Molecular Weight (g/mol) 150.62
MDL Number MFCD00035214
InChI Key WUMMJVLSXMYDRW-UHFFFAOYSA-N
PubChem CID 80240
IUPAC Name hydrogen 4-methyl-1,3-thiazol-2-amine chloride
SMILES [H+].[Cl-].CC1=CSC(N)=N1

Specifications

Melting Point 173°C
Color White
Quantity 25 g
Formula Weight 150.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylthiazole Hydrochloride
EINECSNumber (5)-0203
RTECSNumber XJ2630000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.