Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylthiazole 99.0+%, TCI America™
SDP

Catalog No. A1303250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1303250G 250 g
A130325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1303250G Supplier TCI America Supplier No. A1303250G

Chemical Identifiers

CAS 1603-91-4
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD00005329
InChI Key OUQMXTJYCAJLGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiazole, 4-methylthiazol-2-amine, 2-thiazolamine, 4-methyl, 2-amino-4-methylthiazol, 4-methyl-2-thiazolamine, 4-methyl-2-thiazolylamine, 2-amino-4-methyl-1,3-thiazole, normotiroide, thiazole, 2-amino-4-methyl, 4-methyl-2-aminothiazole
PubChem CID 74143
ChEBI CHEBI:39753
IUPAC Name 4-methyl-1,3-thiazol-2-amine
SMILES CC1=CSC(=N1)N

Specifications

Melting Point 45°C
Color Yellow
Boiling Point 232°C
Quantity 250 g
Formula Weight 114.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylthiazole
EINECSNumber (5)-0203
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.