Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Amino-4-thiazolyl)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. A0954250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0954250G 250 g
A095425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0954250G Supplier TCI America Supplier No. A0954250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29676-71-9
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00010068
InChI Key DYCLHZPOADTVKK-UHFFFAOYSA-N
Synonym 2-amino-4-thiazoleacetic acid, 2-2-aminothiazol-4-yl acetic acid, 2-aminothiazol-4-acetic acid, 2-aminothiazole-4-yl acetic acid, 4-thiazoleacetic acid, 2-amino, 2-amino-1,3-thiazol-4-yl acetic acid, 2-aminothiazole-4-yl-acetic acid, 2-aminothiazole-4-acetic acid, unii-qsh3o4tnjt, 2-2-amino-1,3-thiazol-4-yl acetic acid
PubChem CID 34665
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid
SMILES C1=C(N=C(S1)N)CC(=O)O

Specifications

Melting Point 123°C
Quantity 250 g
Formula Weight 158.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Amino-4-thiazolyl)acetic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.