Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5,6-dimethylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. A146125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A146125G 25 g
A14615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A146125G Supplier TCI America Supplier No. A146125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29927-08-0
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.25
MDL Number MFCD00005790
InChI Key IODWHFFPQHUDAG-UHFFFAOYSA-N
PubChem CID 34757
IUPAC Name 5,6-dimethyl-1,3-benzothiazol-2-amine
SMILES CC1=C(C)C=C2N=C(N)SC2=C1

Specifications

Melting Point 190°C
Quantity 25 g
Synonym 2-amino-5,6-dimethylbenzothiazole, 5,6-dimethylbenzo d thiazol-2-amine, 2-benzothiazolamine, 5,6-dimethyl, 5,6-dimethyl-2-aminobenzothiazole, unii-6ybu8z59ys, 5,6-dimethylbenzothiazol-2-ylamine, 6ybu8z59ys, 5,6-dimethyl-2-benzothiazolamine, 2-benzothiazolamine,5,6-dimethyl, 2-amino-5,6-dimethyl-1,3-benzothiazol
Formula Weight 178.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5,6-dimethylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.