Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromopyridine, 98%

Catalog No. AAA1428014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1428014 25 g
AAA1428006 5 g
AAA1428022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1428014 Supplier Thermo Scientific Chemicals Supplier No. A1428014
Add to Cart

CAS: 1072-97-5 | C5H5BrN2 | 173.013 g/mol

2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.

Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-97-5
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD00006323
InChI Key WGOLHUGPTDEKCF-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyridine, 2-pyridinamine, 5-bromo, 5-bromo-2-pyridylamine, 5-bromo-2-aminopyridine, 5-bromo-2-pyridinamine, 5-bromopyridin-2-ylamine, unii-d5qe8xw52u, 5-bromo-pyridin-2-ylamine, 2-amino-5-bromo pyridine, d5qe8xw52u
PubChem CID 70622
IUPAC Name 5-bromopyridin-2-amine
SMILES C1=CC(=NC=C1Br)N

Specifications

Melting Point 135°C to 140°C
Quantity 25 g
Beilstein 108737
Solubility Information Soluble in methanol,chloroform,ethyl acetate. Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 2-Amino-5-bromopyridine
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-019-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.