Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chloro-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A19311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19311G 1 g
A19315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19311G Supplier TCI America Supplier No. A19311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20712-16-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00129030
InChI Key QTZBTBLHYPSFMG-UHFFFAOYSA-N
Synonym 2-Amino-5-chloro-3-picoline
PubChem CID 350232
IUPAC Name 5-chloro-3-methylpyridin-2-amine
SMILES CC1=CC(Cl)=CN=C1N

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 1 g
Formula Weight 142.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.