Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chlorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. A157225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A157225G 25 g
A15725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A157225G Supplier TCI America Supplier No. A157225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5922-60-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00017106
InChI Key QYRDWARBHMCOAG-UHFFFAOYSA-N
Synonym 5-chloroanthranilonitrile, benzonitrile, 2-amino-5-chloro, anthranilonitrile, 5-chloro, 2-amino-5-chloro-benzonitrile, 4-chloro-2-cyanoaniline, unii-gy0x0082qb, 2-amino-5-chloro benzonitrile, amino 2-5-chloro-benzonitrile, 2-amino-5-chlorobenzenecarbonitrile, acbn
PubChem CID 80019
IUPAC Name 2-amino-5-chlorobenzonitrile
SMILES NC1=CC=C(Cl)C=C1C#N

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Boiling Point 159°C
Formula Weight 152.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Amino-5-chlorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.