Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. A0837100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0837100G 100 g
A083725G 25 g
A0837500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0837100G Supplier TCI America Supplier No. A0837100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1072-98-6
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00006324
InChI Key MAXBVGJEFDMHNV-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
PubChem CID 66174
IUPAC Name 5-chloropyridin-2-amine
SMILES C1=CC(=NC=C1Cl)N

Specifications

Melting Point 136°C
Quantity 100 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.