Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-fluorobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. A115825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A115825G 25 g
A11585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A115825G Supplier TCI America Supplier No. A115825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 393-39-5
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00007831
InChI Key LRCQLCWUUBSUOY-UHFFFAOYSA-N
Synonym 2-amino-5-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl aniline, 4-fluoro-2-trifluoromethyl-phenylamine, benzenamine, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluidine, alpha, 2-amino-5-fluoro benzotrifluoride, 4-fluoro-2-trifluoromethyl phenylamine, alpha,alpha,alpha-4-tetrafluoro-o-toluidine, benzenamine,4-fluoro-2-trifluoromethyl
PubChem CID 67853
IUPAC Name 4-fluoro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(F)C=C1C(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 179.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-5-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.