Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-methyl-3-thiophenecarbonitrile 98.0+%, TCI America™
SDP

Catalog No. A235125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A235125G 25 g
A23515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A235125G Supplier TCI America Supplier No. A235125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138564-58-6
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.188
MDL Number MFCD01626055
InChI Key YGXADLPRHBRTPG-UHFFFAOYSA-N
Synonym 2-Amino-3-cyano-5-methylthiophene
PubChem CID 689056
IUPAC Name 2-amino-5-methylthiophene-3-carbonitrile
SMILES CC1=CC(=C(S1)N)C#N

Specifications

Melting Point 103°C
Quantity 25 g
Formula Weight 138.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methyl-3-thiophenecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.