Learn More
Medchemexpress LLC 2-amino-5-methylthiophene-3-carbonitrile | 138564-58-6 | MFCD01626055 | 99.8% | 138.19 g/mol | C6H6N2S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0157731G
Encompass_Preferred
2-Amino-5-methylthiophene-3-carbonitrile is an organic intermediate used in research and synthetic chemistry. Supplied as a solid with high reported purity, it serves as a building block in heterocyclic synthesis and is applied in routes toward pharmaceutical scaffolds.
  • High reported purity suitable for research applications.
  • Solid form for convenient storage and handling.
  • Useful as a building block in heterocyclic synthesis.
  • Available in multiple pack sizes from milligram to bulk quantities.
  • Analytical documents available (COA, LCMS, SDS) for quality verification.
Catalog No. 50-003-00690
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.