Learn More
Medchemexpress LLC 2-amino-5-methylthiophene-3-carbonitrile | 138564-58-6 | MFCD01626055 | 99.8% | 138.19 | C6H6N2S | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW015773500MG
Encompass_Preferred
2-Amino-5-methylthiophene-3-carbonitrile is a thiophene-derived chemical intermediate for research use, commonly employed as a building block in medicinal chemistry and in synthetic routes toward heterocyclic pharmaceutical scaffolds. The material is provided with analytical characterization to support laboratory workflows.
  • High purity material suitable for research applications.
  • Molecular formula C6H6N2S and molecular weight 138.19.
  • Available as solids and as DMSO solutions for convenient handling.
  • Characterization documents provided, including COA, LC-MS, and SDS.
  • Intended for laboratory research use only; not for human or clinical use.
Catalog No. 50-003-01535