Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-phenyl-1,3,4-thiadiazol 98.0+%, TCI America™
SDP

Catalog No. A30025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30025G 5 g
A30021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30025G Supplier TCI America Supplier No. A30025G

Chemical Identifiers

CAS 2002-03-1
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.23
MDL Number MFCD00205278
InChI Key UHZHEOAEJRHUBW-UHFFFAOYSA-N
Synonym 5-Phenyl-1,3,4-thiadiazol-2-amine
PubChem CID 219408
IUPAC Name 5-phenyl-1,3,4-thiadiazol-2-amine
SMILES C1=CC=C(C=C1)C2=NN=C(S2)N

Specifications

Melting Point 227°C
Quantity 5 g
Formula Weight 177.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-phenyl-1,3,4-thiadiazol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.