Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-chloropurine Riboside 95.0+%, TCI America™
SDP

Catalog No. A20541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20541G 1 g
A20545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20541G Supplier TCI America Supplier No. A20541G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2004-07-1
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005735
InChI Key TXWHPSZYRUHEGT-UHFFFAOYNA-N
Synonym 2-amino-6-chloropurine riboside, 2-amino-6-chloropurine-9-riboside
PubChem CID 46874491
IUPAC Name (3R,4S,5R)-2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2Cl)N

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 1 g
Formula Weight 301.69
Percent Purity ≥95.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloropurine Riboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.