Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-methylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. A0714100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0714100G 100 g
A071425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0714100G Supplier TCI America Supplier No. A0714100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2536-91-6
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005789
InChI Key DZWTXWPRWRLHIL-UHFFFAOYSA-N
Synonym 2-amino-6-methylbenzothiazole, 6-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methyl, 6-methyl-2-aminobenzothiazole, 2-methylbenzothiazol-2-amine, 6-methyl-2-benzothiazolylamine, benzothiazole, 2-amino-6-methyl, 6-methyl-benzothiazol-2-ylamine, 2-amino-6-methyl-benzothiazole, 6-methyl-2-benzothiazolamine
PubChem CID 17335
IUPAC Name 6-methyl-1,3-benzothiazol-2-amine
SMILES CC1=CC=C2N=C(N)SC2=C1

Specifications

Melting Point 138°C
Color Yellow
Quantity 100 g
Formula Weight 164.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methylbenzothiazole
EINECSNumber (5)-0235
RTECSNumber DL2625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.