Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Amino-6-methylheptane

Catalog No. D161292100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D161292100G 100 g
D16129225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D161292100G Supplier Sigma Aldrich Supplier No. D161292100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-amino-6-methylheptane can be used as:
  • A building block for the synthesis of n-type photoconductor, n,nα-bis(1,5-dimethylhexyl)-3,4:9,10-perylenebis (dicarbox-imide) (pdhep)
  • A reagent in the synthesis of substituted pyrrolidones by reductive amination of levulinic acid using supported platinum catalyst
  • A nucleophilic amine in the synthesis of unsymmetrical sulfamide by reacting with sulfamic acid salts and triphenylphosphine ditriflate

Specifications

CAS 543-82-8
Density 0.767 g/mL (at 25°C (literature))
Boiling Point 154°C to 156°C (lit.)
Molecular Formula C8H19N
Refractive Index n20/D 1.422 (literature)
Linear Formula (CH3)2 CH(CH2)3 CH(CH3)NH2
MDL Number MFCD00008092
Quantity 100 g
Synonym 1,5-Dimethylhexylamine; 6-Methyl-2-heptylamine
Molecular Weight (g/mol) 129.24
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.