Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-(trifluoromethyl)benzothiazole 98.0+%, TCI America™
SDP

Catalog No. A24611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24611G 1 g
A24615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24611G Supplier TCI America Supplier No. A24611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 777-12-8
Molecular Formula C8H5F3N2S
Molecular Weight (g/mol) 218.197
MDL Number MFCD00269597
InChI Key WEDYEBJLWMPPOK-UHFFFAOYSA-N
Synonym 2-amino-6-trifluoromethyl benzothiazole, 6-trifluoromethyl benzo d thiazol-2-amine, 6-trifluoromethyl-1,3-benzothiazol-2-amine, 2-amino-6-trifluoromethyl-1,3-benzothiazole, 2-amino-6-trifluoromethylbenzothiazole, 6-trifluoromethyl-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-trifluoromethyl, 2-amino-6-trifluoromethyl-1,3-benzothiazol, 6-trifluoromethyl-1,3-benzothiazol-2-ylamine, 6-trifluoromethyl benzothiazole-2-ylamine
PubChem CID 2735955
IUPAC Name 6-(trifluoromethyl)-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1C(F)(F)F)SC(=N2)N

Specifications

Melting Point 116°C
Color Yellow
Quantity 1 g
Formula Weight 218.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-(trifluoromethyl)benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.