Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. A24351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24351G 1 g
A24355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24351G Supplier TCI America Supplier No. A24351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34486-24-3
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD03093868
InChI Key NFYYDQMFURPHCC-UHFFFAOYSA-N
Synonym 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w
PubChem CID 821024
IUPAC Name 6-(trifluoromethyl)pyridin-2-amine
SMILES C1=CC(=NC(=C1)N)C(F)(F)F

Specifications

Melting Point 87°C
Quantity 1 g
UN Number 2811
Formula Weight 162.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.