Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals

Catalog No. AAH64771MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64771MD 250 mg
AAH6477106 5 g
AAH6477103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64771MD Supplier Thermo Scientific Chemicals Supplier No. H64771MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108714-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD09953790
InChI Key GUTJITRKAMCHSD-UHFFFAOYSA-N
Synonym 2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
PubChem CID 22617093
IUPAC Name 9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C

Specifications

Color White
Quantity 250 mg
Beilstein 15506865
Formula Weight 209.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-9,9-dimethylfluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.