Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Aminoacetanilide 98.0+%, TCI America™
SDP

Catalog No. A242625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A242625G 25 g
A24265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A242625G Supplier TCI America Supplier No. A242625G

Chemical Identifiers

CAS 34801-09-7
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00210388
InChI Key MPXAYYWSDIKNTP-UHFFFAOYSA-N
Synonym n-2-aminophenyl acetamide, 2'-aminoacetanilide, o-aminoacetanilide, acetamide, n-2-aminophenyl, unii-lb34xrq95v, lb34xrq95v, acetamide, n-2-aminophenyl-9ci, n-acetyl-2-aminoaniline, n-acetyl-o-phenylenediamine, 2-aminoactanilide
PubChem CID 11149
IUPAC Name N-(2-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1N

Specifications

Melting Point 134°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Aminoacetanilide
EINECSNumber (3)-0244
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.