Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzimidazole 98.0+%, TCI America™
SDP

Catalog No. A085025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A085025G 25 g
A08505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A085025G Supplier TCI America Supplier No. A085025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 934-32-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005596
InChI Key JWYUFVNJZUSCSM-UHFFFAOYSA-N
Synonym 2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
PubChem CID 13624
ChEBI CHEBI:27822
IUPAC Name 1H-1,3-benzodiazol-2-amine
SMILES NC1=NC2=CC=CC=C2N1

Specifications

Melting Point 231°C
Quantity 25 g
Formula Weight 133.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzimidazole
EINECSNumber (5)-0462
RTECSNumber DD5775000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.