Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzoxazole 98.0+%, TCI America™
SDP

Catalog No. A277425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A277425G 25 g
A27745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A277425G Supplier TCI America Supplier No. A277425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4570-41-6
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00182850
InChI Key JPBLHOJFMBOCAF-UHFFFAOYSA-N
PubChem CID 20707
IUPAC Name 1,3-benzoxazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(O2)N

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Formula Weight 134.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzoxazole
RTECSNumber DM4500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.