Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzylamine 98.0+%, TCI America™
SDP

Catalog No. A143525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A143525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A143525G Supplier TCI America Supplier No. A143525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4403-69-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00075408
InChI Key GVOYKJPMUUJXBS-UHFFFAOYSA-N
Synonym 2-(Aminomethyl)aniline, alpha-Amino-o-toluidine
PubChem CID 178096
IUPAC Name 2-(aminomethyl)aniline
SMILES C1=CC=C(C(=C1)CN)N

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 98°C
Quantity 25 g
UN Number 3259
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.