Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. A212925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A212925G 25 g
A21295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A212925G Supplier TCI America Supplier No. A212925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 534-85-0
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007685
InChI Key NFCPRRWCTNLGSN-UHFFFAOYSA-N
Synonym 2-aminodiphenylamine, n-phenyl-o-phenylenediamine, n1-phenylbenzene-1,2-diamine, o-semidine, 1,2-benzenediamine, n-phenyl, o-aminodiphenylamine, n-phenyl-1,2-phenylenediamine, n-phenylbenzene-1,2-diamine, o-phenylenediamine, n-phenyl, unii-g2x60k1y26
PubChem CID 68297
IUPAC Name N1-phenylbenzene-1,2-diamine
SMILES NC1=CC=CC=C1NC1=CC=CC=C1

Specifications

Melting Point 80°C
Boiling Point 108°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminodiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.