Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethanethiol Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. A0296100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0296100G 100 g
A029625G 25 g
A0296500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0296100G Supplier TCI America Supplier No. A0296100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
SMILES [H+].[Cl-].NCCS

Specifications

Melting Point 68°C
Color White
pH 3.5 to 4.5 (40w/v% H2O soln.)
Quantity 100 g
Formula Weight 113.60
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanethiol Hydrochloride
EINECSNumber (9)-1422
RTECSNumber KJ0200000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.