Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoisobutyric acid, 99%

Catalog No. AAA1302122
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302122 100 g
AAA1302114 25 g
AAA1302130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302122 Supplier Thermo Scientific Chemicals Supplier No. A1302122
Add to Cart

CAS: 62-57-7 | C4H9NO2 | 103.121 g/mol

2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 506496
Merck Index 14,445
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 103.12
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid
EINECSNumber 200-544-0
RTECSNumber AY7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.