Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Aminomethyl)-1-ethylpyrrolidine 98.0+%, TCI America™
SDP

Catalog No. A255625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A255625G 25 g
A25565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A255625G Supplier TCI America Supplier No. A255625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26116-12-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00003178
InChI Key UNRBEYYLYRXYCG-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
PubChem CID 117295
IUPAC Name (1-ethylpyrrolidin-2-yl)methanamine
SMILES CCN1CCCC1CN

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
UN Number 2733
Formula Weight 128.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)-1-ethylpyrrolidine
EINECSNumber (9)-0211
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.