Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenol-4-sulfonamide 97.0+%, TCI America™
SDP

Catalog No. A103125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A103125G 25 g
A1031500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A103125G Supplier TCI America Supplier No. A103125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-32-8
Molecular Formula C6H8N2O3S
Molecular Weight (g/mol) 188.20
MDL Number MFCD00035782
InChI Key AVQFHKYAVVQYQO-UHFFFAOYSA-N
Synonym 2-aminophenol-4-sulfonamide, 4-hydroxymetanilamide, aminophenol sulfamide, benzenesulfonamide, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenesulphonamide, o-aminophenol-p-sulfonamide, 3-amino-4-hydroxy-benzenesulfonamide, metanilamide, 4-hydroxy, 2-aminophenol-4-sulfamide, 3-amino-4-hydroxybenzene-1-sulfonamide
PubChem CID 66814
IUPAC Name 3-amino-4-hydroxybenzene-1-sulfonamide
SMILES NC1=CC(=CC=C1O)S(N)(=O)=O

Specifications

Melting Point 199°C
Quantity 25 g
Formula Weight 188.20
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenol-4-sulfonamide
EINECSNumber (3)-2030
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.