Learn More
2-Aminophenol, 99%
Description
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
Solubility
Soluble in water (17 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 173°C to 177°C |
| Density | 1.328 |
| Boiling Point | 150°C (11 mmHg) |
| Flash Point | >175°C (347°F) |
| Odor | Characteristic |
| Quantity | 2500 g |
| UN Number | UN2512 |
| Beilstein | 606075 |
| Merck Index | 14,461 |
| Solubility Information | Soluble in water (17g/L at 20°C). |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H317-H341 |
| Hazard Statement | GHS H Statement H341-H302-H332 Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-) |
| EINECSNumber | 202-431-1 |
| RTECSNumber | SJ4950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.