Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenol 98.0+%, TCI America™
SDP

Catalog No. A0335100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0335100G 100 g
A033525G 25 g
A0335500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0335100G Supplier TCI America Supplier No. A0335100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O

Specifications

Melting Point 175°C
Quantity 100 g
UN Number 2512
Formula Weight 109.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenol
EINECSNumber (3)-0675
RTECSNumber SJ4950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.