Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminopyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. A07781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07781G 1 g
A0778100MG 100 mg
A07785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A07781G Supplier TCI America Supplier No. A07781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14150-95-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00059168
InChI Key KTPMVZCGIJJWCD-UHFFFAOYSA-N
PubChem CID 139694
IUPAC Name 2-imino-1,2-dihydropyridin-1-ol
SMILES ON1C=CC=CC1=N

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.