Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amyl-5-methylphenol 98.0+%, TCI America™
SDP

Catalog No. A27101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27101G 1 g
A27105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27101G Supplier TCI America Supplier No. A27101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1300-94-3
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00127710
InChI Key CKGWFZQGEQJZIL-UHFFFAOYSA-N
Synonym 5-Methyl-2-pentylphenol
PubChem CID 14759
ChEBI CHEBI:48213
IUPAC Name 5-methyl-2-pentylphenol
SMILES CCCCCC1=C(C=C(C=C1)C)O

Specifications

Boiling Point 139°C
Quantity 1 g
UN Number 3145
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amyl-5-methylphenol
EINECSNumber (3)-0521
RTECSNumber GP3100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.