Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Anilino-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. A27411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27411G 1 g
A27415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27411G Supplier TCI America Supplier No. A27411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 355832-04-1
Molecular Formula C21H19N
Molecular Weight (g/mol) 285.39
MDL Number MFCD20040462
InChI Key RFTRFDMRINNTSI-UHFFFAOYSA-N
PubChem CID 18730177
IUPAC Name 9,9-dimethyl-N-phenylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)NC4=CC=CC=C4)C

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 1 g
Formula Weight 285.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Anilino-9,9-dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.