Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Anthraceneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A1843250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1843250MG 250 mg
A18431G 1 g
A1843-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1843250MG Supplier TCI America Supplier No. A1843250MG

Chemical Identifiers

CAS 141981-64-8
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD07784001
InChI Key PKWBMOXZIMVOJT-UHFFFAOYSA-N
PubChem CID 9855952
IUPAC Name (anthracen-2-yl)boronic acid
SMILES OB(O)C1=CC=C2C=C3C=CC=CC3=CC2=C1

Specifications

Quantity 250 mg
Formula Weight 222.05
Physical Form Crystalline Powder
Chemical Name or Material 2-Anthraceneboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.