Learn More
Apexbio Technology LLC 2-APB(Synonyms: 2-Aminoethoxydiphenyl borate, 2-Aminoethyl diphenylborinate, 2-Aminoethoxydiphenylborane), 10mM (in 1mL DMSO), CAS: 524-95-8.
SDP
Supplier:  Apexbio Technology LLC B66435.1
Encompass_Preferred
2-APB (CAS 524-95-8) is a cell-permeable antagonist of Ins(1 4 5)P3-induced calcium release It inhibits calcium mobilization mediated by the intracellular IP3 receptor (IP3R) an essential component in Ca2 signaling events such as calcium oscillations and waves In rat cerebellar microsomes 2-APB inhibits Ins(1 4 5)P3-induced Ca2 release with an IC50 of 42 M Additionally it blocks TRPC channels including TRPC3 TRPC5 (IC50 20 M in HEK-293 cells) and TRPC6 2-APB is widely utilized in research focusing on intracellular calcium dynamics channel modulation studies and oxidative stress-related cell injury
Catalog No. 50-115-2604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.