Learn More
Cayman Chemical 2-Arachidonoyl Glycerol, 53847-30-6, 1 mg
SDP
Supplier:  Cayman Chemical 621601
Encompass_Preferred
An endogenous CB1 and CB2 receptor agonist (Kis = 25.3-472 and 145-1,400 nM, respectively); the most abundant molecular species of MAG found in rat brain; formation is calcium-dependent and is mediated by the activities of PLC and DAG lipase; induces a rapid, transient increase in intracellular free calcium in NG108-15 neuroblastoma X glioma cells through a CB1 receptor-dependent mechanism at a concentration of 0.3 nM; metabolized in vitro by MAG lipase and FAAH, with MAG lipase likely being the principle metabolizing enzyme in vivo,
Catalog No. NC9740619
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.