Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzoylpyrrole 98.0+%, TCI America™
SDP

Catalog No. B493425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B493425G 25 g
B49345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B493425G Supplier TCI America Supplier No. B493425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7697-46-3
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.199
MDL Number MFCD00094048
InChI Key NFGGQMYSOLVBLF-UHFFFAOYSA-N
PubChem CID 24370
IUPAC Name phenyl(1H-pyrrol-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CN2

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 305°C
Quantity 25 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylpyrrole
RTECSNumber OB6407000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.