Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone 98.0+%, TCI America™
SDP

Catalog No. B3914100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3914100G 100 g
B391425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3914100G Supplier TCI America Supplier No. B3914100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119313-12-1
Molecular Formula C23H30N2O2
Molecular Weight (g/mol) 366.505
MDL Number MFCD00191775
InChI Key UHFFVFAKEGKNAQ-UHFFFAOYSA-N
Synonym 2-Benzyl-2-(dimethylamino)-1-[4-(morpholino)phenyl]-1-butanone
PubChem CID 86171
IUPAC Name 2-benzyl-2-(dimethylamino)-1-(4-morpholin-4-ylphenyl)butan-1-one
SMILES CCC(CC1=CC=CC=C1)(C(=O)C2=CC=C(C=C2)N3CCOCC3)N(C)C

Specifications

Melting Point 116°C
Color Yellow
Quantity 100 g
UN Number 3077
Formula Weight 366.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyl-2-(dimethylamino)-4′-morpholinobutyrophenone
EINECSNumber (5)-6310
RTECSNumber EL7755000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.