Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B476625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B476625G 25 g
B47665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B476625G Supplier TCI America Supplier No. B476625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14389-86-7
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00051940
InChI Key GMOYUTKNPLBTMT-UHFFFAOYSA-M
Synonym 2-benzyloxy benzoic acid, 2-benzyloxybenzoic acid, benzoic acid,2-phenylmethoxy, 2-phenylmethoxy benzoic acid, benzoic acid, 2-phenylmethoxy, enamine_005339, benzyloxybenzoic acid, acmc-20ao3m, 2-benzyloxy-benzoic acid, 2-benzyloxybenzoicacid
PubChem CID 1810581
IUPAC Name 2-(benzyloxy)benzoate
SMILES [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 228.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.