Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC440320500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440320500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440320500 Supplier Thermo Scientific Chemicals Supplier No. 440320500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328000-16-4 , 109-99-9
Molecular Formula C13H11BrMgO
Molecular Weight (g/mol) 287.44
MDL Number MFCD08705262
InChI Key VELKJPFZDYXBDE-UHFFFAOYSA-M
Synonym 2-benzyloxyphenylmagnesium bromide solution, velkjpfzdyxbde-uhfffaoysa-m, 2-benzyloxyphenyl magnesium bromide, 2-benzyloxy phenylmagnesium bromide, 2-benzyloxy phenyl magnesium bromide, 2-benzyloxyphenylmagnesium bromide, 1m, 2-benzyloxy phenyl bromo magnesium, 2-benzyloxyphenylmagnesium bromide, 1m, magnesium, bromo 2-phenylmethoxy phenyl, grignard reagent
PubChem CID 13183249
IUPAC Name magnesium;phenylmethoxybenzene;bromide
SMILES Br[Mg]C1=CC=CC=C1OCC1=CC=CC=C1

Specifications

CAS Min % 26.08, 69.8
CAS Max % 30.2, 73.92
Color Brown to Yellow
Density 1.0470g/mL
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Linear Formula C6H5CH2OC6H4
Quantity 50 mL
Specific Gravity 1.047
Formula Weight 287.43
Physical Form Solution
Chemical Name or Material 2-Benzyloxyphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.