Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Biphenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. I050325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I050325G 25 g
I05035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I050325G Supplier TCI America Supplier No. I050325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17337-13-2
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00037087
InChI Key IHHUGFJSEJSCGE-UHFFFAOYSA-N
Synonym 2-biphenylyl isocyanate, 2-biphenylisocyanate, 2-biphenyl isocyanate, 2-isocyanatobiphenyl, 2-isocyanato-biphenyl, 2-biphenylylisocyanate, isocyanic acid 2-biphenyl ester, 1,1'-biphenyl, isocyanato, 1-isocyanato-2-phenyl-menzene, 1,1-biphenyl-2-yl isocyanate
PubChem CID 222280
IUPAC Name 2-isocyanato-1,1'-biphenyl
SMILES O=C=NC1=CC=CC=C1C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 2206
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Biphenyl Isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.