Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B010025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B010025G 25 g
B01005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B010025G Supplier TCI America Supplier No. B010025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-92-5
Molecular Formula C20H18O3
Molecular Weight (g/mol) 306.36
MDL Number MFCD00059144
InChI Key CREICILGVGNQBH-UHFFFAOYSA-N
Synonym Phenolphthalol
PubChem CID 66495
IUPAC Name 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol
SMILES OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 204°C
Color White
Quantity 25 g
Formula Weight 306.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol
RTECSNumber DA4912000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.