Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Boc-amino)ethanol, 95%
Click to view available options
Quantity:
5 g
25 g
Description
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.
Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.
Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Boiling Point | 92°C (0.22mmHg) |
Refractive Index | 1.449 |
Linear Formula | (CH3)3CCO2NHCH2CH2OH |
Solubility Information | Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol. |
Density | 1.042 |
Quantity | 25 g |
Formula Weight | 161.2 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Boc-amino)ethanol |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction