Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. B44561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44561G 1 g
B44565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44561G Supplier TCI America Supplier No. B44561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22426-14-8
Molecular Formula C12H7BrN2
Molecular Weight (g/mol) 259.106
MDL Number MFCD08276287
InChI Key ZRJUDAZGVGIDLP-UHFFFAOYSA-N
PubChem CID 12830247
IUPAC Name 2-bromo-1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=C2)C=CC(=N3)Br)N=C1

Specifications

Melting Point 163°C
Quantity 1 g
Formula Weight 259.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.